Assay Details
| Gene Symbol: AT1G35250 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001333146.1 | NP_001320228.1 | 1 - 2 | 332 | 99 | |
| NM_001333147.1 | NP_001320227.1 | 1 - 2 | 330 | 99 | ||
| NM_103223.4 | NP_174759.1 | 1 - 2 | 332 | 99 | ||
| GenBank mRNA | BT024833.1 | - | 1 - 2 | 141 | 99 | |
More Information
Set Membership: |
|

AT1G35250
Genomic Map