Assay Details
| Gene Symbol: AT1G35290 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_103226.3 | NP_564457.1 | 2 - 3 | 313 | 154 | |
| GenBank mRNA | AF428452.1 | - | 2 - 3 | 304 | 154 | |
| AY125550.1 | - | 2 - 3 | 242 | 154 | ||
More Information
Set Membership: |
|

AT1G35290
Genomic Map