Assay Details
| Gene Symbol: RanBPM | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_103241.3 | NP_174777.2 | 4 - 5 | 765 | 137 | |
| NM_202234.3 | NP_973963.1 | 4 - 5 | 844 | 137 | ||
| GenBank mRNA | AY070090.1 | - | 4 - 5 | 765 | 137 | |
| AY096710.1 | - | 4 - 5 | 673 | 137 | ||
More Information
Set Membership: |
|

RanBPM
Genomic Map