Assay Details
| Gene Symbol: RanBPM | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_103241.3 | NP_174777.2 | 5 - 6 | 845 | 112 | |
| NM_202234.3 | NP_973963.1 | 5 - 6 | 924 | 112 | ||
| GenBank mRNA | AY070090.1 | - | 5 - 6 | 845 | 112 | |
| AY096710.1 | - | 5 - 6 | 753 | 112 | ||
More Information
Set Membership: |
|

RanBPM
Genomic Map