Assay Details
| Gene Symbol: PDIL5-2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_103262.5 | NP_564462.1 | 2 - 3 | 472 | 90 | |
| GenBank mRNA | AF386986.1 | - | 2 - 3 | 346 | 90 | |
| BT008751.1 | - | 2 - 3 | 314 | 90 | ||
More Information
Set Membership: |
|

PDIL5-2
Genomic Map