Assay Details
| Gene Symbol: ANNAT1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_103274.4 | NP_174810.1 | 1 - 2 | 207 | 88 | |
| GenBank mRNA | AF083913.1 | - | 1 - 2 | 104 | 88 | |
| AF332435.1 | - | 1 - 2 | 71 | 88 | ||
| AY072347.1 | - | 1 - 2 | 103 | 88 | ||
| AY086570.1 | - | 1 - 2 | 106 | 88 | ||
| BT003359.1 | - | 1 - 2 | 71 | 88 | ||
More Information
Set Membership: |
|

ANNAT1
Genomic Map