Assay Details
| Gene Symbol: ACC1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001198214.2 | NP_001185143.1 | 20 - 21 | 2926 | 91 | |
| NM_103313.4 | NP_174849.2 | 20 - 21 | 2897 | 91 | ||
| GenBank mRNA | D34630.2 | - | 19 - 20 | 2536 | 91 | |
More Information
Set Membership: |
|

ACC1
Genomic Map