Assay Details
| Gene Symbol: AT1G37000 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_103355.1 | NP_174892.1 | 2 - 3 | 151 | 96 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

AT1G37000| Gene Symbol: AT1G37000 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_103355.1 | NP_174892.1 | 2 - 3 | 151 | 96 | |
Set Membership: |
|