Assay Details
| Gene Symbol: UGT80B1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001084205.2 | NP_001077674.1 | 4 - 5 | 721 | 105 | |
| NM_001333213.1 | NP_001319161.1 | 4 - 5 | 655 | 105 | ||
| NM_001333214.1 | NP_001320890.1 | 2 - 3 | 1018 | 105 | ||
| NM_001333215.1 | NP_001320888.1 | 3 - 4 | 934 | 105 | ||
| NM_001333216.1 | NP_001320891.1 | 3 - 4 | 739 | 105 | ||
| NM_103487.4 | NP_175027.1 | 3 - 4 | 805 | 105 | ||
| NM_202238.3 | NP_973967.1 | 3 - 4 | 558 | 105 | ||
| GenBank mRNA | AK227397.1 | - | 3 - 4 | 678 | 105 | |
| BT005834.1 | - | 2 - 3 | 292 | 105 | ||
| KJ138970.1 | - | 2 - 3 | 292 | 105 | ||
More Information
Set Membership: |
|

UGT80B1
Genomic Map