Assay Details
| Gene Symbol: UGT80B1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001084205.2 | NP_001077674.1 | 10 - 11 | 1555 | 65 | |
| NM_001333213.1 | NP_001319161.1 | 10 - 11 | 1489 | 65 | ||
| NM_001333214.1 | NP_001320890.1 | 8 - 9 | 1852 | 65 | ||
| NM_001333215.1 | NP_001320888.1 | 9 - 10 | 1768 | 65 | ||
| NM_001333216.1 | NP_001320891.1 | 9 - 10 | 1573 | 65 | ||
| NM_001333217.1 | NP_001320889.1 | 5 - 6 | 718 | 65 | ||
| NM_103487.4 | NP_175027.1 | 9 - 10 | 1639 | 65 | ||
| NM_202238.3 | NP_973967.1 | 9 - 10 | 1392 | 65 | ||
| GenBank mRNA | AK227397.1 | - | 9 - 10 | 1512 | 65 | |
| BT005834.1 | - | 8 - 9 | 1126 | 65 | ||
| KJ138970.1 | - | 8 - 9 | 1126 | 65 | ||
More Information
Set Membership: |
|

UGT80B1
Genomic Map