Assay Details
| Gene Symbol: RAB18 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001084206.1 | NP_001077675.1 | 6 - 7 | 639 | 73 | |
| NM_001123963.1 | NP_001117435.1 | 5 - 6 | 645 | 73 | ||
| NM_103516.4 | NP_175056.1 | 6 - 7 | 675 | 73 | ||
| GenBank mRNA | AF370263.1 | - | 6 - 7 | 599 | 73 | |
| AK227664.1 | - | 6 - 7 | 634 | 73 | ||
| AY063080.1 | - | 5 - 6 | 528 | 73 | ||
| AY086878.1 | - | 6 - 7 | 638 | 73 | ||
| U75603.1 | - | 5 - 6 | 547 | 73 | ||
More Information
Set Membership: |
|

RAB18
Genomic Map