Assay Details
| Gene Symbol: CYCA1;1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_103537.5 | NP_175077.1 | 5 - 6 | 1168 | 64 | |
| GenBank mRNA | AK175406.1 | - | 5 - 6 | 1063 | 64 | |
More Information
Set Membership: |
|

CYCA1;1
Genomic Map