Assay Details
| Gene Symbol: CYCA1;1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_103537.5 | NP_175077.1 | 7 - 8 | 1493 | 86 | |
| GenBank mRNA | AK175406.1 | - | 7 - 8 | 1388 | 86 | |
More Information
Set Membership: |
|

CYCA1;1
Genomic Map