Assay Details
| Gene Symbol: AT1G44224 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001084209.2 | NP_001077678.1 | 1 | 272 | 65 | |
| GenBank mRNA | AK220577.1 | - | 1 | 257 | 65 | |
More Information
Set Membership: |
|

AT1G44224
Genomic Map