Assay Details
| Gene Symbol: CH1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_103548.5 | NP_175088.1 | 1 - 2 | 203 | 83 | |
| NM_202240.2 | NP_973969.1 | 1 - 2 | 156 | 83 | ||
| NM_202241.2 | NP_973970.1 | 1 - 2 | 156 | 83 | ||
| GenBank mRNA | AK228645.1 | - | 1 - 2 | 121 | 83 | |
| AY128357.1 | - | 1 - 2 | 156 | 83 | ||
| BT002075.1 | - | 1 - 2 | 84 | 83 | ||
More Information
Set Membership: |
|

CH1
Genomic Map