Assay Details
| Gene Symbol: CH1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001333230.1 | NP_001321683.1 | 1 - 2 | 306 | 86 | |
| NM_103548.5 | NP_175088.1 | 2 - 3 | 439 | 86 | ||
| NM_202240.2 | NP_973969.1 | 2 - 3 | 392 | 86 | ||
| NM_202241.2 | NP_973970.1 | 2 - 3 | 392 | 86 | ||
| GenBank mRNA | AK228645.1 | - | 2 - 3 | 357 | 86 | |
| AY128357.1 | - | 2 - 3 | 392 | 86 | ||
| BT002075.1 | - | 2 - 3 | 320 | 86 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001333230.1 transcripts.
Set Membership: |
|

CH1
Genomic Map