Assay Details
| Gene Symbol: SIAR1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_103561.4 | NP_175101.1 | 6 - 7 | 1115 | 134 | |
| GenBank mRNA | AY049306.1 | - | 6 - 7 | 1056 | 134 | |
| AY149964.1 | - | 6 - 7 | 920 | 134 | ||
More Information
Set Membership: |
|

SIAR1
Genomic Map