Assay Details
| Gene Symbol: AT1G44970 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_103577.4 | NP_175117.1 | 3 - 4 | 773 | 74 | |
| GenBank mRNA | AK228469.1 | - | 3 - 4 | 650 | 74 | |
| BT008612.1 | - | 3 - 4 | 650 | 74 | ||
| X98804.1 | - | 637 | 74 | |||
More Information
Set Membership: |
|

AT1G44970
Genomic Map