Assay Details
| Gene Symbol: SBP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_103601.3 | NP_175141.1 | 2 - 3 | 196 | 68 | |
| GenBank mRNA | AF324720.1 | - | 2 - 3 | 160 | 68 | |
| AF329502.1 | - | 2 - 3 | 159 | 68 | ||
| AF349529.1 | - | 2 - 3 | 139 | 68 | ||
| AK221737.1 | - | 2 - 3 | 185 | 68 | ||
| AY070470.1 | - | 2 - 3 | 159 | 68 | ||
| AY093744.1 | - | 2 - 3 | 139 | 68 | ||
More Information
Set Membership: |
|

SBP1
Genomic Map