Assay Details
Gene Symbol: RD21A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_103612.3 | NP_564497.1 | 4 - 5 | 1443 | 87 | |
GenBank mRNA | AY072130.1 | - | 4 - 5 | 1363 | 87 | |
AY133781.1 | - | 4 - 5 | 1303 | 87 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
