Assay Details
| Gene Symbol: GAMMA CA2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_103620.4 | NP_175159.1 | 1 - 2 | 258 | 156 | |
| GenBank mRNA | AY045979.1 | - | 1 - 2 | 227 | 156 | |
| AY113936.1 | - | 1 - 2 | 121 | 156 | ||
More Information
Set Membership: |
|

GAMMA CA2
Genomic Map