Assay Details
| Gene Symbol: 3BETAHSD/D1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_103623.5 | NP_564502.1 | 5 - 6 | 776 | 136 | |
| NM_179448.4 | NP_849779.1 | 5 - 6 | 874 | 136 | ||
| GenBank mRNA | AK117478.1 | - | 5 - 6 | 771 | 136 | |
| BT005166.1 | - | 4 - 5 | 633 | 136 | ||
More Information
Set Membership: |
|

3BETAHSD/D1
Genomic Map