Assay Details
| Gene Symbol: AT1G47670 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_103660.6 | NP_175198.1 | 1 - 2 | 614 | 76 | |
| GenBank mRNA | AY065445.1 | - | 1 - 2 | 427 | 76 | |
| AY117276.1 | - | 1 - 2 | 381 | 76 | ||
More Information
Set Membership: |
|

AT1G47670
Genomic Map