Assay Details
| Gene Symbol: MYB112 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_103696.3 | NP_564519.1 | 1 - 2 | 275 | 70 | |
| GenBank mRNA | AK229083.1 | - | 1 - 2 | 246 | 70 | |
| AY008377.2 | - | 1 - 2 | 275 | 70 | ||
| AY519562.1 | - | 1 - 2 | 192 | 70 | ||
More Information
Set Membership: |
|

MYB112
Genomic Map