Assay Details
| Gene Symbol: CHR18 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001333322.1 | NP_001321270.1 | 13 - 14 | 1240 | 116 | |
| NM_001333323.1 | NP_001321271.1 | 13 - 14 | 1117 | 116 | ||
| NM_001333324.1 | NP_001321269.1 | 13 - 14 | 1219 | 116 | ||
| NM_103728.5 | NP_175265.3 | 13 - 14 | 1225 | 116 | ||
More Information
Set Membership: |
|

CHR18
Genomic Map