Assay Details
| Gene Symbol: CHR18 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001333323.1 | NP_001321271.1 | 2 - 3 | 263 | 99 | |
| NM_001333324.1 | NP_001321269.1 | 2 - 3 | 365 | 99 | ||
| NM_103728.5 | NP_175265.3 | 2 - 3 | 371 | 99 | ||
More Information
Set Membership: |
|

CHR18
Genomic Map