Assay Details
| Gene Symbol: D-CDES | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001333334.1 | NP_001319174.1 | 3 - 4 | 551 | 130 | |
| NM_001333335.1 | NP_001320840.1 | 3 - 4 | 545 | 130 | ||
| NM_103738.7 | NP_175275.3 | 3 - 4 | 542 | 130 | ||
More Information
Set Membership: |
|

D-CDES
Genomic Map