Assay Details
| Gene Symbol: JAZ4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001123978.2 | NP_001117450.1 | 2 - 3 | 511 | 69 | |
| NM_001123979.2 | NP_001117451.1 | 1 - 2 | 547 | 69 | ||
| NM_103746.3 | NP_175283.2 | 2 - 3 | 511 | 69 | ||
| GenBank mRNA | AY954762.1 | - | 2 - 3 | 243 | 69 | |
| DQ132675.1 | - | 2 - 3 | 425 | 69 | ||
More Information
Set Membership: |
|

JAZ4
Genomic Map