Assay Details
| Gene Symbol: JAZ4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001123978.2 | NP_001117450.1 | 5 - 6 | 1037 | 65 | |
| NM_001123979.2 | NP_001117451.1 | 4 - 5 | 1073 | 65 | ||
| NM_103746.3 | NP_175283.2 | 5 - 6 | 1037 | 65 | ||
| GenBank mRNA | AY954762.1 | - | 5 - 6 | 769 | 65 | |
| DQ132675.1 | - | 5 - 6 | 951 | 65 | ||
More Information
Set Membership: |
|

JAZ4
Genomic Map