Assay Details
| Gene Symbol: JAZ4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_103746.3 | NP_175283.2 | 6 - 7 | 1089 | 119 | |
| GenBank mRNA | AY954762.1 | - | 6 - 7 | 821 | 119 | |
| DQ132675.1 | - | 6 - 7 | 1003 | 119 | ||
More Information
Set Membership: |
|

JAZ4
Genomic Map