Assay Details
| Gene Symbol: AT1G48570 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_103753.5 | NP_175290.1 | 3 - 4 | 975 | 75 | |
| GenBank mRNA | AY065170.1 | - | 3 - 4 | 975 | 75 | |
| BT000067.1 | - | 3 - 4 | 917 | 75 | ||
More Information
Set Membership: |
|

AT1G48570
Genomic Map