Assay Details
| Gene Symbol: AT3G57900 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_115652.1 | NP_191349.1 | 1 - 2 | 100 | 80 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

AT3G57900| Gene Symbol: AT3G57900 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_115652.1 | NP_191349.1 | 1 - 2 | 100 | 80 | |
Set Membership: |
|