Assay Details
| Gene Symbol: AT3G58200 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_115682.4 | NP_191379.1 | 2 - 3 | 427 | 98 | |
| GenBank mRNA | BT004114.1 | - | 2 - 3 | 363 | 98 | |
| BT005114.1 | - | 2 - 3 | 269 | 98 | ||
More Information
Set Membership: |
|

AT3G58200
Genomic Map