Assay Details
| Gene Symbol: AT3G58260 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_115688.2 | NP_191385.1 | 2 - 3 | 529 | 99 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

AT3G58260| Gene Symbol: AT3G58260 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_115688.2 | NP_191385.1 | 2 - 3 | 529 | 99 | |
Set Membership: |
|