Assay Details
| Gene Symbol: AT3G58380 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_115700.3 | NP_191397.1 | 2 - 3 | 292 | 141 | |
| GenBank mRNA | AK176427.1 | - | 2 - 3 | 252 | 141 | |
| BT010442.1 | - | 1 - 2 | 106 | 141 | ||
More Information
Set Membership: |
|

AT3G58380
Genomic Map