Assay Details
| Gene Symbol: RBL15 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001203197.1 | NP_001190126.1 | 12 - 13 | 1254 | 73 | |
| NM_115708.4 | NP_567064.1 | 11 - 12 | 1387 | 73 | ||
| GenBank mRNA | AY136386.1 | - | 11 - 12 | 1221 | 73 | |
| BT000186.1 | - | 11 - 12 | 1126 | 73 | ||
More Information
Set Membership: |
|

RBL15
Genomic Map