Assay Details
| Gene Symbol: RBL15 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001203197.1 | NP_001190126.1 | 10 - 11 | 1160 | 91 | |
| NM_115708.4 | NP_567064.1 | 9 - 10 | 1293 | 91 | ||
| GenBank mRNA | AY136386.1 | - | 9 - 10 | 1127 | 91 | |
| BT000186.1 | - | 9 - 10 | 1032 | 91 | ||
More Information
Set Membership: |
|

RBL15
Genomic Map