Assay Details
| Gene Symbol: PRN | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_115784.3 | NP_191481.1 | 1 - 2 | 492 | 65 | |
| GenBank mRNA | AF353716.1 | - | 1 - 2 | 170 | 65 | |
| BT026056.1 | - | 1 - 2 | 99 | 65 | ||
More Information
Set Membership: |
|

PRN
Genomic Map