Assay Details
| Gene Symbol: TXR1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001339956.1 | NP_001327906.1 | 3 - 4 | 434 | 93 | |
| NM_115790.3 | NP_567078.1 | 3 - 4 | 418 | 93 | ||
| GenBank mRNA | AF380643.1 | - | 3 - 4 | 418 | 93 | |
| AY054136.1 | - | 2 - 3 | 215 | 93 | ||
More Information
Set Membership: |
|

TXR1
Genomic Map