Assay Details
| Gene Symbol: FTA | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_115800.6 | NP_567084.1 | 3 - 4 | 498 | 115 | |
| GenBank mRNA | BT033079.1 | - | 3 - 4 | 459 | 115 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

FTA| Gene Symbol: FTA | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_115800.6 | NP_567084.1 | 3 - 4 | 498 | 115 | |
| GenBank mRNA | BT033079.1 | - | 3 - 4 | 459 | 115 | |
Set Membership: |
|