Assay Details
| Gene Symbol: GCN2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001203206.1 | NP_001190135.1 | 20 - 21 | 3180 | 70 | |
| NM_001339969.1 | NP_001326041.1 | 20 - 21 | 4047 | 70 | ||
| NM_001339970.1 | NP_001326042.1 | 19 - 20 | 3189 | 70 | ||
| NM_001339971.1 | NP_001326040.1 | 19 - 20 | 2960 | 70 | ||
| NM_115803.3 | NP_191500.2 | 19 - 20 | 3079 | 70 | ||
| GenBank mRNA | AJ459823.1 | - | 19 - 20 | 2727 | 70 | |
More Information
Set Membership: |
|

GCN2
Genomic Map