Assay Details
| Gene Symbol: AT3G59480 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_115810.4 | NP_191507.1 | 2 - 3 | 853 | 105 | |
| GenBank mRNA | BT028928.1 | - | 2 - 3 | 739 | 105 | |
More Information
Set Membership: |
|

AT3G59480
Genomic Map