Assay Details
| Gene Symbol: AT3G59580 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001035818.3 | NP_001030895.1 | 2 - 3 | 1140 | 72 | |
| NM_001339989.1 | NP_001327143.1 | 3 - 4 | 968 | 72 | ||
| NM_115820.4 | NP_191517.1 | 2 - 3 | 967 | 72 | ||
More Information
Set Membership: |
|

AT3G59580
Genomic Map