Assay Details
| Gene Symbol: AT3G59580 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001035818.3 | NP_001030895.1 | 4 - 5 | 2036 | 87 | |
| NM_001339989.1 | NP_001327143.1 | 5 - 6 | 1864 | 87 | ||
| NM_115820.4 | NP_191517.1 | 4 - 5 | 1863 | 87 | ||
More Information
Set Membership: |
|

AT3G59580
Genomic Map