Assay Details
| Gene Symbol: LSM6A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001340000.1 | NP_001327468.1 | 2 - 3 | 560 | 81 | |
| NM_001340001.1 | NP_001327469.1 | 3 - 4 | 387 | 81 | ||
| NM_115843.3 | NP_191540.1 | 3 - 4 | 375 | 81 | ||
| GenBank mRNA | BT002850.1 | - | 3 - 4 | 316 | 81 | |
| BT004433.1 | - | 2 - 3 | 222 | 81 | ||
More Information
Set Membership: |
|

LSM6A
Genomic Map