Assay Details
| Gene Symbol: BGLU16 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001035822.2 | NP_001030899.1 | 7 - 8 | 956 | 64 | |
| NM_001084847.1 | NP_001078316.1 | 7 - 8 | 1144 | 64 | ||
| NM_115876.5 | NP_191572.1 | 8 - 9 | 1423 | 64 | ||
| GenBank mRNA | AY045953.1 | - | 8 - 9 | 1147 | 64 | |
| AY113935.1 | - | 8 - 9 | 1106 | 64 | ||
More Information
Set Membership: |
|

BGLU16
Genomic Map