Assay Details
| Gene Symbol: DL1E | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_115882.4 | NP_567094.1 | 1 - 2 | 473 | 145 | |
| GenBank mRNA | AF428332.1 | - | 1 - 2 | 295 | 145 | |
| AJ304842.1 | - | 1 - 2 | 255 | 145 | ||
More Information
Set Membership: |
|

DL1E
Genomic Map