Assay Details
| Gene Symbol: EIF4G | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001084848.1 | NP_001078317.1 | 1 - 2 | 316 | 82 | |
| NM_001084849.2 | NP_001078318.1 | 1 - 2 | 392 | 82 | ||
| NM_001084850.1 | NP_001078319.1 | 1 - 2 | 390 | 82 | ||
| GenBank mRNA | AB107249.1 | - | 1 - 2 | 316 | 82 | |
More Information
Set Membership: |
|

EIF4G
Genomic Map