Assay Details
| Gene Symbol: EDA14 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_115900.4 | NP_191595.1 | 2 - 3 | 279 | 65 | |
| GenBank mRNA | AY039876.1 | - | 2 - 3 | 251 | 65 | |
| AY085156.1 | - | 2 - 3 | 234 | 65 | ||
| AY101514.1 | - | 1 - 2 | 60 | 65 | ||
| BT000725.1 | - | 2 - 3 | 195 | 65 | ||
More Information
Set Membership: |
|

EDA14
Genomic Map