Assay Details
| Gene Symbol: EDA14 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_115900.4 | NP_191595.1 | 7 - 8 | 708 | 98 | |
| GenBank mRNA | AY039876.1 | - | 7 - 8 | 680 | 98 | |
| AY085156.1 | - | 7 - 8 | 663 | 98 | ||
| AY101514.1 | - | 6 - 7 | 489 | 98 | ||
| BT000725.1 | - | 7 - 8 | 624 | 98 | ||
More Information
Set Membership: |
|

EDA14
Genomic Map